Skip to Content
Merck
All Photos(1)

Key Documents

37-0280

Sigma-Aldrich

Zinc chloride

JIS special grade, ≥98.0%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor pressure

1 mmHg ( 428 °C)

Assay

≥98.0%

form

solid

reaction suitability

reagent type: catalyst
core: zinc

availability

available only in Japan

pH

5 (20 °C, 100 g/L)

mp

293 °C (lit.)

storage temp.

15-25°C

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Davies et al.
Cell death & disease, 2, e136-e136 (2011-04-01)
The activation and regulation of target genes by the tumour-suppressor p53 dictates the fate of a cell, with cell cycle arrest or apoptosis being two distinct outcomes. PERP (p53 apoptosis effector related to PMP-22), a p53 transcriptional target, is induced
Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were
İbrahim Gülseren et al.
Food chemistry, 135(2), 770-774 (2012-08-08)
Whey protein isolate (WPI) nanoparticles were prepared using ethanol desolvation, and their capacity to incorporate ZnCl(2) was analysed. Desolvation was carried out at pH 9 and the volume of added ethanol was 0-3 times the volume of protein solution. The
Michele K Sankako et al.
Pharmacological reports : PR, 64(6), 1537-1546 (2013-02-15)
The aim of this study was to evaluate the changes in testicular function of rats due to cigarette smoke exposure and the possible mechanism by which zinc protects against these alterations. Male Wistar rats (60 days old) were randomly divided
Hatice Yorulmaz et al.
Biological trace element research, 151(2), 256-262 (2012-11-28)
We evaluated the effect of zinc treatment on the blood-brain barrier (BBB) permeability and the levels of zinc (Zn), natrium (Na), magnesium (Mg), and copper (Cu) in the brain tissue during epileptic seizures. The Wistar albino rats were divided into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service