Skip to Content
Merck
All Photos(1)

Key Documents

14422

Sigma-Aldrich

Zinc chloride

puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1 mmHg ( 428 °C)

Quality Level

grade

puriss.

Assay

98-100.5%

form

crystalline powder

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

heavy metals, complies
oxide chloride (as ZnO), complies
residual solvents, complies
≤0.005% foreign heavy metals (as Pb)
≤0.005% total nitrogen (N)
≤0.03% ammonium (NH4)
≤1% alkalis/earth alkalis

pH

4.6-5.5 (20 °C, 10%)

mp

293 °C (lit.)

anion traces

sulfate (SO42-): ≤200 mg/kg

cation traces

Al:, in accordance
As: ≤2 mg/kg
Ca: ≤100 mg/kg
Ca:, in accordance
Fe: ≤10 mg/kg
Fe:, in accordance
K: ≤200 mg/kg
Mg: ≤100 mg/kg
Mg:, in accordance
Na: ≤100 mg/kg

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride is a zinc salt. Zinc chloride in combination with 1-ethyl-3-methylimidazolium chloride comprises a molten salt, which is employed in the electrodeposition studies of zinc.
Zinc chloride is a colorless white compound composed of zinc and chlorine, which can be used as a flux for soldering.

Application

Zinc chloride may be employed as catalyst in Knoevenagel condensation of carbonyl compounds with active methylene group containing reagents. It may be used in preparation of porous carbon nanofibers, useful in the fabrication of efficient electrodes for supercapacitors. It may be used as catalyst in the preparation of poly(propylene fumarate).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AIEEE Chemistry null
Albert K Shung et al.
Journal of biomaterials science. Polymer edition, 13(1), 95-108 (2002-05-11)
Diethyl fumarate and propylene glycol were reacted in the presence of a zinc chloride catalyst to synthesize poly(propylene fumarate) (PPF) over a period of 12 hours. The kinetics of the transesterification polymerization at 130 degrees C, 150 degrees C, and
Self-Sustained Thin Webs Consisting of Porous Carbon Nanofibers for Supercapacitors via the Electrospinning of Polyacrylonitrile Solutions Containing Zinc
Kim C, et al.
Advanced Mat., 19(17), 2341-2346 (2007)
Electrodeposition of zinc from a Lewis acidic zinc chloride-1-ethyl-3-methylimidazolium chloride molten salt.
Lin Y-F and Sun I.
Electrochimica Acta, 44(16), 2771-2777 (1999)
Zinc chloride as a new catalyst for Knoevenagel condensation.
Shanthan RP and Venkataratnam RV.
Tetrahedron Letters, 32(41), 5821-5822 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service