Skip to Content
Merck
All Photos(1)

Key Documents

450111

Sigma-Aldrich

Zinc chloride

AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
synthesis grade
form:
amorphous
solubility:
H2O: soluble

grade

synthesis grade

Quality Level

vapor pressure

1 mmHg ( 428 °C)

product line

AnhydroBeads

Assay

99.99% trace metals basis

form

amorphous

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤150.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

293 °C (lit.)

solubility

H2O: soluble

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in the low temperature synthesis of nanocrystalline zinc oxide films. Also used in the synthesis of zinc oxide nanoparticles with low agglomeration. Aqueous suspensions of the nanoparticles displayed high transmittance in the visible light range, but exhibited strong absorption in the UV range.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuzuki, T., et al.
J. Autralasian Ceram. Soc., 38, 15-15 (2002)
Gorbik, P.P., et al.
Dopovidi Natsional'noi Akademii Nauk Ukranaie, 1, 135-135 (2004)
Andrea M Hui et al.
Dermatologic clinics, 30(3), 503-515 (2012-07-18)
For melanoma treatment, the primary goal is complete removal with histologically negative margins. Mohs micrographic surgery (MMS) has been extensively used and studied for the treatment of nonmelanoma skin cancer, particularly at sites where tissue conservation is vital. The use
Manuella P Kaster et al.
Pharmacological reports : PR, 64(3), 706-713 (2012-07-21)
In this work, the contribution of NMDA receptors to the antidepressant-like effect of adenosine in the forced swimming test (FST) was investigated. The pretreatment of mice with NMDA or D-serine was able to prevent the anti-immobility effect of either adenosine
L Davies et al.
Cell death & disease, 2, e136-e136 (2011-04-01)
The activation and regulation of target genes by the tumour-suppressor p53 dictates the fate of a cell, with cell cycle arrest or apoptosis being two distinct outcomes. PERP (p53 apoptosis effector related to PMP-22), a p53 transcriptional target, is induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service