This is a dual-layer SPE tube containing both Supelclean ENVI-Carb (upper layer) and LC-NH2 (lower layer) SPE sorbents, separated by a polyethylene frit. The ENVI-Carb layer is a carbon surface with a hexagonal ring structure. The LC-NH2 layer is an aminopropyl (5%) monomerically bonded onto carbon.
54035-U
Supelclean™ ENVI-Carb/NH2 SPE Tube
bed A 500 mg (Supelclean ENVI-Carb), bed B 500 mg (Supelclean LC-NH2), volume 6 mL, pk of 30
Synonym(s):
Carbon/NH2 SPE cartidge, 6 mL
About This Item
Recommended Products
material
polypropylene tube
product line
Supelclean™
composition
bed A, 500 mg (Supelclean ENVI-Carb)
bed B, 500 mg (Supelclean LC-NH2)
packaging
pk of 30
technique(s)
solid phase extraction (SPE): suitable
volume
6 mL
matrix active group
aminopropyl phase
carbon phase
application(s)
food and beverages
separation technique
ion exchange
reversed phase
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Organic or aqueous solutions
- Dual layer SPE tube that contains both Supelclean ENVI-Carb (upper layer) & LC-NH2 (lower layer) SPE sorbents (separated by PE frit)
- Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.).
- ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products
- Supelclean LC-NH2 is an aminopropyl phase that retains fatty acids, organic acids, and some polar pigments and sugars common in food matrices
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is the Supelclean ENVI-Carb/NH2 SPE Tube Column an activated carbon column as a classification? Or is it a GC/NH2 column?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service