Skip to Content
Merck
All Photos(1)

Documents

54067-U

Supelco

Supelclean ENVI-Carb II/PSA SPE Tube

bed B 500 mg (Supelclean PSA SPE), bed A 500 mg (Supeclean ENVI-Carb-II SPE), volume 6 mL, pk of 30

Synonym(s):

Carbon/PSA SPE cartidge, 6 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

PE frit (20 μm porosity)
polypropylene hardware

product line

Supelclean

composition

bed A, 500 mg (Supeclean ENVI-Carb-II SPE)
bed B, 500 mg (Supelclean PSA SPE)

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

matrix active group

PSA phase
carbon phase

application(s)

food and beverages

separation technique

ion exchange
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Reversed-phase and anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • Dual layer SPE tube that contains both Supelclean ENVI-Carb (upper layer) & PSA (lower layer) SPE sorbents (separated by PE frit)
  • Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.).
  • ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products
  • Supelclean PSA is a polymerically bonded, ethylenediamine-N-propyl phase that contains both primary and secondary amines
  • Supelclean PSA has a strong affinity and high capacity for fatty acids, organic acids, and some polar pigments and sugars
  • Tested for superior cleanliness using GC-FID and GC-MS

Legal Information

ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service