28017-U
Diffusion Caps
Standard
Synonym(s):
Diffusion Cap, Standard, 10 pk
About This Item
Recommended Products
manufacturer/tradename
L4070207
maximum temperature
100 °C
technique(s)
active air sampling: suitable
H × W × D
17 mm × 10.0 mm × 15 mm
application(s)
air monitoring
environmental
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service