Skip to Content
Merck
All Photos(3)

Key Documents

T8877

Sigma-Aldrich

2,3,5-Triphenyltetrazolium chloride

≥98.0% (HPLC)

Synonym(s):

TPTZ, TTZ, Tetrazolium Red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15ClN4
CAS Number:
Molecular Weight:
334.80
Beilstein:
3923356
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

≥98.0% (HPLC)

form

powder

color

faintly yellow

solubility

H2O: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].c1ccc(cc1)-c2nn(-c3ccccc3)[n+](n2)-c4ccccc4

InChI

1S/C19H15N4.ClH/c1-4-10-16(11-5-1)19-20-22(17-12-6-2-7-13-17)23(21-19)18-14-8-3-9-15-18;/h1-15H;1H/q+1;/p-1

InChI key

PKDBCJSWQUOKDO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5-Triphenyltetrazolium chloride is a colorless water-soluble dye. In the mitochondria of living cells, it is reduced to a deep red, water-insoluble compound (formazan). 2,3,5-Triphenyltetrazolium chloride helps to distinguish between viable and infarcted brain tissue after stroke.

Application

2,3,5-Triphenyltetrazolium chloride has been used to stain heart tissue to measure the extent of acute lesion. It has also been used to stain brain tissue to check the size of infarct area.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chesarahmia Dojo Soeandy et al.
Apoptosis : an international journal on programmed cell death, 24(7-8), 578-595 (2019-05-11)
Over the past 30 years a number of animal models of cerebral ischemic injury have been developed. Middle cerebral artery occlusion (MCAO) in particular reproduces both ischemic and reperfusion elements and is widely utilized as a model of ischemic stroke in
Evaluation of a novel epicardial atrial fibrillation treatment system.
Kiser AC et al.
The Annals of Thoracic Surgery, 85, 300-300 (2008)
Human umbilical mesenchymal stem cells promote recovery after ischemic stroke.
Lin YC et al.
Stroke, 42, 2045-2045 (2011)
Mannitol-facilitated perfusion staining with 2,3,5-triphenyltetrazolium chloride (TTC) for detection of experimental cerebral infarction and biochemical analysis.
Sun YY et al.
Journal of Neuroscience Methods, 203, 122-122 (2012)
Jia Huang et al.
Journal of molecular and cellular cardiology, 134, 107-118 (2019-07-14)
Myocardial ischemia/reperfusion (I/R) injury is the major limitation for the cardioprotective action of revascularization after myocardial infarction. Lymphocyte adapter protein (Lnk), an adapter protein, has a regulatory role in multiple signaling pathways by functioning as a scaffold for different substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service