Skip to Content
Merck
All Photos(1)

Documents

W1628

Sigma-Aldrich

Wortmannin

from Penicillium funiculosum, ≥98% (HPLC and TLC)

Synonym(s):

KY 12420

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O8
CAS Number:
Molecular Weight:
428.43
Beilstein:
67676
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

biological source

Penicillium funiculosum

Quality Level

Assay

≥98% (HPLC and TLC)

form

powder

color

off-white

solubility

DMSO: soluble
H2O: unstable

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC(=O)[C@@]1(C)C[C@@H](OC(C)=O)C3=C2C(=O)c4occ5C(=O)O[C@H](COC)[C@@]3(C)c45

InChI

1S/C23H24O8/c1-10(24)30-13-7-22(2)12(5-6-14(22)25)16-18(13)23(3)15(9-28-4)31-21(27)11-8-29-20(17(11)23)19(16)26/h8,12-13,15H,5-7,9H2,1-4H3/t12-,13+,15+,22-,23-/m0/s1

InChI key

QDLHCMPXEPAAMD-QAIWCSMKSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Wortmannin is a potent and specific phosphatidylinositol 3-kinase (PI3-K) inhibitor with an IC50 of 2-4 nM. Inhibition of PI3-K/Akt signal transduction cascade enhances the apoptotic effects of radiation or serum withdrawal and blocks the antiapoptotic effect of cytokines. Inhibition of PI3-K by wortmannin also blocks many of the short-term metabolic effects induced by insulin receptor activation.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide and Kinase Phosphatase Biology research. Discover more featured Cyclic Nucleotide and Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the G Proteins (Heterotrimeric) and Phosphoinositide Kinases pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Acute Tox. 2 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nakanishi et al.
The Journal of biological chemistry, 267(4), 2157-2163 (1992-02-05)
We have found that a fungal strain, Talaromyces wortmannin KY12420, produces a potent inhibitor of smooth muscle myosin light chain kinase (MLCK). This active product, designated as MS-54, was isolated and purified from the culture broth of the fungus and
Alban R Pereira et al.
Journal of medicinal chemistry, 53(24), 8523-8533 (2010-12-03)
Analogues of the sponge meroterpenoid liphagal have been synthesized and evaluated for inhibition of PI3Kα and PI3Kγ as part of a program aimed at developing new isoform-selective PI3K inhibitors. One of the analogues, compound 24, with IC₅₀ values of 66
Alexander D Crawford et al.
PloS one, 6(2), e14694-e14694 (2011-03-08)
Natural products represent a significant reservoir of unexplored chemical diversity for early-stage drug discovery. The identification of lead compounds of natural origin would benefit from therapeutically relevant bioassays capable of facilitating the isolation of bioactive molecules from multi-constituent extracts. Towards
Matthew G K Benesch et al.
Journal of lipid research, 56(6), 1134-1144 (2015-04-22)
Autotaxin (ATX) is a secreted enzyme, which produces extracellular lysophosphatidate (LPA) from lysophosphatidylcholine (LPC). LPA activates six G protein-coupled receptors and this is essential for vasculogenesis during embryonic development. ATX is also involved in wound healing and inflammation, and in
M Chekenya et al.
Oncogene, 27(39), 5182-5194 (2008-05-13)
Chemoresistance represents a major problem in the treatment of many malignancies. Overcoming this obstacle will require improved understanding of the mechanisms responsible for this phenomenon. The progenitor cell marker NG2/melanoma proteoglycan (MPG) is aberrantly expressed by various tumors, but its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service