Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,3,5-Triphenyltetrazolium chloride
Synonym(s):
TPTZ, TTZ, Tetrazolium Red, 2,3,5-Triphenyltetrazolium chloride
Empirical Formula (Hill Notation):
C
19
H
15
ClN
4
CAS Number:
298-96-4
Molecular Weight:
334.80
EC Number:
206-071-6
Products
Product Category
media supplements (2)
building blocks (1)
stains (1)
Brand
Millipore (1)
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Special Grade
Food Grade (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T8877
2,3,5-Triphenyltetrazolium chloride,
≥98.0% (HPLC)
Expand
View Pricing
1.08380
2,3,5-Triphenyltetrazolium chloride,
Color indicator. Supplement in culture media for the microbiological analysis of food and water samples
Expand
View Pricing
30-4890
2,3,5-Triphenyltetrazolium chloride,
JIS special grade, 98.0-102.0%
Expand
View Pricing