Skip to Content
Merck
All Photos(2)

Documents

D2650

Sigma-Aldrich

Dimethyl sulfoxide

Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.75

grade

Hybri-Max

Quality Level

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

sterility

sterile-filtered

product line

BioReagent

Assay

≥99.7%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

cell culture | hybridoma: suitable
cryopreservation: suitable

impurities

endotoxin, tested
≤0.1% water
water

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

solubility

H2O: miscible (completely)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions (PCR) and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines; embryonic stem cells (ESC), and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum (FBS).

Packaging

100 mL in amber bottle
Glass ampoule packaging is now available under a new product number, D2653

Other Notes

This product is a Hybri-Max product. It is hybridoma tested and is assessed for suitability in cell freezing. This product is sterile filtered (0.2 micron) and tested for endotoxin levels.

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azadeh Ghaee et al.
Materials science & engineering. C, Materials for biological applications, 103, 109767-109767 (2019-07-28)
In this study, a simple one-step method was introduced to prepare suitable PCL wound dressing by using a small amount of hydrophilic surface modifying macromolecule (LSMM) with ends capped with Polyethylene glycol and aminated zeolitic imidazolate frameworks (MZIF-8) as a
Stepan Coufal et al.
Cells, 8(7) (2019-07-25)
Crohn's disease (CD), ulcerative colitis (UC) and inflammatory bowel disease (IBD) associated with primary sclerosing cholangitis (PSC-IBD), share three major pathogenetic mechanisms of inflammatory bowel disease (IBD)-gut dysbiosis, gut barrier failure and immune system dysregulation. While clinical differences among them
Hongxiu Diao et al.
BMC veterinary research, 15(1), 276-276 (2019-08-04)
Mammary gland tumor is the most common spontaneous tumor in intact female dogs, and its poor prognosis remains a clinical challenge. Ivermectin, a well-known anti-parasitic agent, has been implicated as a potential anticancer agent in various types of human cancer.
Likun Gao et al.
Cell death & disease, 10(8), 609-609 (2019-08-14)
Paeonol (Pae), a phenolic acid compound isolated from the Moutan Cortex, was previously demonstrated to exert multiple anticancer effects. The rational control of autophagy has been considered a potential treatment strategy for epithelial ovarian cancer. However, whether Pae induces autophagy
Shady Sayed et al.
Methods (San Diego, Calif.), 164-165, 36-48 (2019-05-13)
The CRISPR/Cas9 system is transforming many biomedical disciplines, including cancer research. Through its flexible programmability and efficiency to induce DNA double strand breaks it has become straightforward to introduce cancer mutations into cells in vitro and/or in vivo. However, not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service