Skip to Content
Merck
All Photos(2)

Key Documents

R109

Sigma-Aldrich

Ro 15-4513

solid

Synonym(s):

Ethyl 8-azido-6-dihydro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Ro15-4513

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N6O3
CAS Number:
Molecular Weight:
326.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

tan

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.8 mg/mL
H2O: insoluble
methanol: soluble

originator

Roche

SMILES string

CCOC(=O)c1ncn-2c1CN(C)C(=O)c3cc(ccc-23)N=[N+]=[N-]

InChI

1S/C15H14N6O3/c1-3-24-15(23)13-12-7-20(2)14(22)10-6-9(18-19-16)4-5-11(10)21(12)8-17-13/h4-6,8H,3,7H2,1-2H3

InChI key

CFSOJZTUTOQNIA-UHFFFAOYSA-N

Application

Ro 15-4513 has been used as a benzodiazepine (Bz) inverse agonist in rats and to test its binding with diazepam-sensitive (DS) receptors in in vitro radioligand binding studies.

Biochem/physiol Actions

Ro 15-4513 is benzodiazepine receptor partial inverse agonist. It elicits anxiogenic property and is an ethanol antagonist. It elicits a high affinity towards diazepam sensitive and insensitive sites in the cerebellar region. Ro 15-4513 possesses an imidazo ring and is an imidazobenzodiazepine.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franco R Mir et al.
British journal of pharmacology, 177(13), 3075-3090 (2020-03-07)
GABAA receptor functions are dependent on subunit composition, and, through their activation, GABA can exert trophic actions in immature neurons. Although several sex differences in GABA-mediated responses are known to be dependent on gonadal hormones, few studies have dealt with
William T Ralvenius et al.
Nature communications, 6, 6803-6803 (2015-04-14)
Agonists at the benzodiazepine-binding site of GABAA receptors (BDZs) enhance synaptic inhibition through four subtypes (α1, α2, α3 and α5) of GABAA receptors (GABAAR). When applied to the spinal cord, they alleviate pathological pain; however, insufficient efficacy after systemic administration
P D Suzdak et al.
Science (New York, N.Y.), 234(4781), 1243-1247 (1986-12-05)
Ethanol, at pharmacologically relevant concentrations of 20 to 100 mM, stimulates gamma-aminobutyric (GABA) receptor-mediated uptake of 36Cl-labeled chlorine into isolated brain vesicles. One drug that acts at GABA-benzodiazepine receptors, the imidazobenzodiazepine Ro15-4513, has been found to be a potent antagonist
David J Nutt et al.
Neuropharmacology, 53(7), 810-820 (2007-09-25)
Alcohol produces many subjective and objective effects in man including pleasure, sedation, anxiolysis, plus impaired eye movements and memory. In human volunteers we have used a newly available GABA-A/benzodiazepine receptor inverse agonist that is selective for the alpha5 subtype (a5IA)
Gian Marco Leggio et al.
Pharmacological research, 141, 384-391 (2019-01-17)
The dopamine D3 receptor (D3R), in the nucleus accumbens (NAc), plays an important role in alcohol reward mechanisms. The major neuronal type within the NAc is the GABAergic medium spiny neuron (MSN), whose activity is regulated by dopaminergic inputs. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service