Skip to Content
Merck
All Photos(2)

Key Documents

85707

Supelco

2,5-Dihydroxybenzoic acid

matrix substance for MALDI-MS, >99.0% (HPLC)

Synonym(s):

2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2209119
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Assay

>99.0% (HPLC)

form

powder

analyte functional class(es)

polymers

analyte chemical class(es)

glycans, lipids, organic molecules, peptides, proteins

technique(s)

MALDI-MS: suitable

mp

200-207 °C
204-208 °C (lit.)

cation traces

Ba: ≤5 mg/kg
Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
K: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

ε (extinction coefficient)

>2000 at 337 nm

suitability

suitable for matrix substance for MALDI-MS

SMILES string

OC(=O)c1cc(O)ccc1O

InChI

1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

WXTMDXOMEHJXQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Dihydroxybenzoic acid is an ionic liquid matrix used for the analysis of biomolecules using matrix-assisted laser desorption/ionization combined with mass spectrometry.
Most commonly used matrix for carbohydrates. For MALDI-MS of free neutral glycans. Produces [M+Na]+ ion; may show a weaker [M+K]+ ion. Other species may be generated by the addition of the appropriate inorganic salt.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix-assisted laser desorption/ionization mass spectrometry with additives to 2,5-di-hydroxy-benzoic acid.
Karas, M., et al.,
Org. Mass Spectrom., 28, 1476-1481 (1993)
Naoki Sato
Plant & cell physiology, 56(10), 1890-1899 (2015-08-16)
Monogalactosyldiacylglycerol (MGDG) is ubiquitous in the photosynthetic membranes of cyanobacteria and chloroplasts. It is synthesized by galactosylation of diacylglycerol (DAG) in the chloroplasts, whereas it is produced by epimerization of monoglucosyldiacylglycerol (GlcDG) in at least several cyanobacteria that have been
2,5-Dihydroxybenzoic acid butylamine and other ionic liquid matrixes for enhanced MALDI-MS analysis of biomolecules
Mank M, et al.
Analytical Chemistry, 76(10), 2938-2950 (2004)
F Bochner et al.
Clinical pharmacology and therapeutics, 30(2), 266-275 (1981-08-01)
Single oral doses of aspirin (ASA, 1,500 mg), sodium salicylate (NaSA, 1,500 mg, 1,200 mg), and salicyluric acid (SUA, 500 mg) were given to five subjects. Serial plasma and urine samples were collected for 24 hr (plasma) and up to
Houjiang Zhou et al.
Molecular & cellular proteomics : MCP, 10(10), M110-M110 (2011-07-01)
Metal and metal oxide chelating-based phosphopeptide enrichment technologies provide powerful tools for the in-depth profiling of phosphoproteomes. One weakness inherent to current enrichment strategies is poor binding of phosphopeptides containing multiple basic residues. The problem is exacerbated when strong cation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service