Skip to Content
Merck
All Photos(1)

Key Documents

I5879

Sigma-Aldrich

3-Isobutyl-1-methylxanthine

≥99% (HPLC), powder

Synonym(s):

1-Methyl-3-isobutylxanthine, 3,7-Dihydro-1-methyl-3-(2-methylpropyl)-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-2,6(1H,3H)-purinedione, IBMX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N4O2
CAS Number:
Molecular Weight:
222.24
Beilstein:
247859
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (HPLC)

form

powder

color

white to off-white

mp

200-201 °C (lit.)

solubility

DMSO: 1 M (with gentle warming)
ethanol: 10 mg/mL

storage temp.

−20°C

SMILES string

CC(C)CN1C(=O)N(C)C(=O)c2[nH]cnc12

InChI

1S/C10H14N4O2/c1-6(2)4-14-8-7(11-5-12-8)9(15)13(3)10(14)16/h5-6H,4H2,1-3H3,(H,11,12)

InChI key

APIXJSLKIYYUKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-isobutyl-1-methylxanthine/IBMX is a non-selective, non-specific inhibitor of cAMP and cGMP phosphodiesterases. IBMX can induce melanogenesis, and can be used as a positive control in melanogenesis research. In oocyte research, IBMX assists in maintaining the germinal vesicle (GV) arrest of prophase I oocytes.

Application

3-Isobutyl-1-methylxanthine has been used:
  • in the differentiation of mesenchymal stem cells (MSC)
  • as a culture medium supplement for inducing adipogenic differentiation
  • as a supplement in Krebs-Ringer buffer with HEPES (KRBH) solution for inducing glucose-stimulated insulin secretion (GSIS)
  • in the characterization of adipose-derived mesenchymal stem cells
  • in cyclic adenosine monophosphate (cAMP) assay
  • in phosphodiesterase (PDE) inhibition

Biochem/physiol Actions

The increase in cAMP level as a result of phosphodiesterase inhibition by IBMX activates PKA, leading to decreased proliferation, increased differentiation, and induction of apoptosis. IBMX inhibits phenylephrine-induced release of 5-hydroxytryptamine from neuroendocrine epithelial cells of the airway mucosa (IC50: 1.3 μM). IBMX also serves as an adenosine receptor antagonist. IBMX has been shown to inhibit ion channels in the neuromuscular junction, GH3 cells, and vascular smooth muscle cells. IBMX induces calcium release from intracellular stores in sensory neurons.
The increase in cAMP level as a result of phosphodiesterase inhibition by IBMX activates PKA, leading to decreased proliferation, increased differentiation, and induction of apoptosis. IBMX inhibits phenylephrine-induced release of 5-hydroxytryptamine from neuroendocrine epithelial cells of the airway mucosa (IC50: 1.3 μM). IBMX also serves as an adenosine receptor antagonist. IBMX has been shown to inhibit ion channels in the neuromuscular junction, GH3 cells, and vascular smooth muscle cells. IBMX induces calcium release from intracellular stores in sensory neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Skin pigmentation enhancers
Comprehensive Series in Photosciences, 3, 637-675 (2001)
Crystal structures of phosphodiesterases 4 and 5 in complex with inhibitor 3-isobutyl-1-methylxanthine suggest a conformation determinant of inhibitor selectivity
Huai Q, et al.
The Journal of Biological Sciences, 279(13), 13095-13101 (2004)
A conserved aromatic lock for the tryptophan rotameric switch in TM-VI of seven-transmembrane receptors
Holst B, et al.
The Journal of Biological Chemistry, 285(6), 3973-3985 (2010)
Culturing INS-1 cells on CDPGYIGSR-, RGD-and fibronectin surfaces improves insulin secretion and cell proliferation.
Kuehn C, et al.
Acta Biomaterialia, 8(2), 619-626 (2012)
Adult bone marrow is a rich source of human mesenchymal `stem?cells but umbilical cord and mobilized adult blood are not
Wexler SA, et al.
British Journal of Haematology, 121(2), 368-374 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service