Skip to Content
Merck
All Photos(1)

Documents

L2387

Sigma-Aldrich

Lipopolysaccharides from Salmonella typhosa

purified by gel-filtration chromatography

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bacterial (Salmonella typhosa)

Quality Level

form

lyophilized powder

purified by

gel-filtration chromatography

impurities

<1% Protein

color

white to faint yellow

solubility

water: soluble

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is extracted from Salmonella typhosa and purified by gel filtration. The source strain is ATCC 10749.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malick M Gibani et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 68(8), 1265-1273 (2018-09-27)
Shedding of Salmonella Typhi or Paratyphi in the stool or urine leads to contamination of food or water, which is a prerequisite for transmission of enteric fever. Currently, there are limited data on the effect of vaccination or prior exposure
Claire S Waddington et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 58(9), 1230-1240 (2014-02-13)
Typhoid fever is a major global health problem, the control of which is hindered by lack of a suitable animal model in which to study Salmonella Typhi infection. Until 1974, a human challenge model advanced understanding of typhoid and was
Thomas C Darton et al.
Frontiers in microbiology, 8, 1794-1794 (2017-10-04)
Current diagnostic tests for typhoid fever, the disease caused by
Thomas C Darton et al.
PLoS neglected tropical diseases, 10(8), e0004926-e0004926 (2016-08-18)
Typhoid persists as a major cause of global morbidity. While several licensed vaccines to prevent typhoid are available, they are of only moderate efficacy and unsuitable for use in children less than two years of age. Development of new efficacious
Malick M Gibani et al.
PLoS neglected tropical diseases, 14(10), e0008783-e0008783 (2020-10-21)
Enteric fever is a systemic infection caused by Salmonella Typhi or Paratyphi A. In many endemic areas, these serovars co-circulate and can cause multiple infection-episodes in childhood. Prior exposure is thought to confer partial, but incomplete, protection against subsequent attacks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service