Skip to Content
Merck
All Photos(1)

Documents

L7895

Sigma-Aldrich

Lipopolysaccharides from Salmonella typhosa

γ-irradiated, BioXtra, suitable for cell culture

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.75

biological source

bacterial (Salmonella typhosa)

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

purified by

gel-filtration chromatography

technique(s)

cell culture | mammalian: suitable

impurities

<1% Protein (Lowry)

color

white to faint yellow

solubility

water: 0.90-1.10 mg/mL, faintly hazy to hazy, colorless to faintly yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lipopolysaccharides from Salmonella typhosa are used in cell culture and cell binding applications. The role of LPS in the secretion of TNF-α and IL-6 was studied in mouse spleen cell culture. It was used to study the role of inflammatory responses in multiple sclerosis.
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne L Jones et al.
Brain : a journal of neurology, 133(Pt 8), 2232-2247 (2010-07-28)
Treatment of early relapsing-remitting multiple sclerosis with the lymphocyte-depleting humanized monoclonal antibody alemtuzumab (Campath [registered trade mark]) significantly reduced the risk of relapse and accumulation of disability compared with interferon β-1a in a phase 2 trial [Coles et al., (Alemtuzumab
Yasuhisa Ano et al.
Nutrients, 11(9) (2019-09-12)
The number of patients with mental illnesses is rapidly increasing, and daily lifestyle is closely associated with the development of symptoms. It is suggested that inflammatory molecules derived from microglia play crucial roles for the pathophysiology of depression. In the
Yasuhisa Ano et al.
Scientific reports, 10(1), 20028-20028 (2020-11-20)
The prevention of age-related cognitive decline and dementia is becoming a high priority because of the rapid growth of aging populations. We have previously shown that hop bitter acids such as iso-α-acids (IAAs) and matured hop bitter acids (MHBAs) activate
Yasuhisa Ano et al.
Molecules (Basel, Switzerland), 24(3) (2019-01-31)
Inflammation in the brain is associated with various disorders including Alzheimer's disease and depression. Thus, inflammation has received increasing attention regarding preventive approaches to such disorders. Epidemiological investigations have reported that drinking tea reduces the risk of dementia and depression.
Aurélien Godinat et al.
Current protocols in chemical biology, 6(3), 169-189 (2014-09-11)
The great complexity of many human pathologies, such as cancer, diabetes, and neurodegenerative diseases, requires new tools for studies of biological processes on the whole organism level. The discovery of novel biocompatible reactions has tremendously advanced our understanding of basic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service