Skip to Content
Merck
All Photos(4)

Documents

L3012

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O111:B4

purified by gel-filtration chromatography

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (O111:B4)

Quality Level

form

lyophilized powder

purified by

gel-filtration chromatography

impurities

<1% Protein

color

white to faint yellow

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is extracted from E. coli serotype O111:B4 and purified by gel filtration. The source strain is from a private collection. This LPS serotype has been used to stimulate B-cells and induce NOS in human hepatocytes.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Huang et al.
Bone research, 5, 17019-17019 (2017-12-22)
The activation of M1 macrophages can be achieved by stimulating them with lipopolysaccharide (LPS) and interferon-γ (IFN-γ). However, M1 can be found under physiological conditions without any pathological stimuli. This study aimed to understand the involvement of RANKL-induced M1 macrophages
Matthew G Frank et al.
Brain, behavior, and immunity, 73, 352-363 (2018-05-29)
Exposure to stressors induces anxiety- and depressive-like behaviors, which are mediated, in part, by neuroinflammatory processes. Recent findings demonstrate that treatment with the immunoregulatory and anti-inflammatory bacterium, Mycobacterium vaccae (M. vaccae), attenuates stress-induced exaggeration of peripheral inflammation and stress-induced anxiety-like
Zhongjie Li et al.
PloS one, 9(5), e97539-e97539 (2014-05-16)
Antimicrobial peptides have attracted much interest as a novel class of antibiotics against a variety of microbes including antibiotics resistant strains. In this study, a new cationic antimicrobial peptide Hp1404 was identified from the scorpion Heterometrus petersii, which is an
Norihiko Takeda et al.
Genes & development, 24(5), 491-501 (2010-03-03)
Hypoxic response and inflammation both involve the action of the hypoxia-inducible transcription factors HIF-1alpha and HIF-2alpha. Previous studies have revealed that both HIF-alpha proteins are in a number of aspects similarly regulated post-translationally. However, the functional interrelationship of these two
Susanne A Wolf et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(9), 3121-3128 (2009-05-13)
To understand the link between peripheral immune activation and neuronal precursor biology, we investigated the effect of T-cell activation on adult hippocampal neurogenesis in female C57Bl/6 mice. A peripheral adaptive immune response triggered by adjuvant-induced rheumatoid arthritis (2 microg/microl methylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service