Skip to Content
Merck
All Photos(2)

Key Documents

L6143

Sigma-Aldrich

Lipopolysaccharides from Salmonella enterica serotype typhimurium

BioXtra, suitable for cell culture, γ-irradiated

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.75

biological source

Salmonella enterica (serotype typhimurium)

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

purified by

gel-filtration chromatography

technique(s)

cell culture | mammalian: suitable

impurities

<1% Protein (Lowry)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).
Lipopolysaccharides from Salmonella enterica serotype typhimurium has been used:
  • in mice to elicit the secretion of pro-inflammatory cytokines
  • to induce cytokine production in monocytes
  • to elicit the activation of NF-κB pathway in mouse dendritic cells.
  • to induce inflammatory response in human colon adenocarcinoma Caco-2 cells
  • for stimulation of the catfish peripheral blood leukocytes (PBLs) to study estrogen receptor mRNA expression

Biochem/physiol Actions

LPS is a major constituent of the cell wall of most gram negative bacteria. It is a highly immunogenic antigen with the ability to enhance immune responses to soluble antigens. LPS also acts as a specific mitogen for bone marrow derived B lymphocytes from mice, rabbits, chickens, cows, hamsters, and humans.

Reconstitution

Lipopolysaccharides are supplied as lyophilized, γ-irradiated powders. To reconstitute, add 1 ml sterile balanced salt solution or tissue culture medium to the vial (1 mg) and gently swirl until the powder dissolves. Reconstituted product may be further diluted to desired working concentrations using sterile balanced salt solution or tissue culture medium.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangwei Xu et al.
Journal of enzyme inhibition and medicinal chemistry, 33(1), 1089-1094 (2018-06-23)
Indoleamine 2,3-dioxygenase 1 (IDO1) activity links to immune escape of cancers. Inhibition of IDO1 provides a new approach for cancer treatment. Most clinical IDO1 drugs show marginal efficacy as single agents. On basis of molecular docking and pharmacophore modelling, a
Luke R Iwanowicz et al.
Fish & shellfish immunology, 40(1), 109-119 (2014-06-29)
Estrogens are recognized as modulators of immune responses in mammals and teleosts. While it is known that the effects of estrogens are mediated via leukocyte-specific estrogen receptors (ERs) in humans and mice, leucocyte-specific estrogen receptor expression and the effects of
Chun-Yan Yeung et al.
Gastroenterology research and practice, 2013, 973209-973209 (2013-07-11)
Background. Lactobacillus shows beneficial anti-inflammatory effects on Salmonella infection. The maintenance of the tight junction (TJ) integrity plays an importance role in avoiding bacterial invasion. Whether Lactobacillus could be used to regulate the TJ protein expression and distribution in inflamed
Sara Artigas-Jerónimo et al.
Vaccines, 8(1) (2020-02-13)
The main objective of this study was to propose a novel methodology to approach challenges in molecular biology. Akirin/Subolesin (AKR/SUB) are vaccine protective antigens and are a model for the study of the interactome due to its conserved function in
Ju Huang et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(15), 6226-6231 (2009-04-03)
Autophagy plays an important role in immunity to microbial pathogens. The autophagy system can target bacteria in phagosomes, promoting phagosome maturation and preventing pathogen escape into the cytosol. Recently, Toll-like receptor (TLR) signaling from phagosomes was found to initiate their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service