Skip to Content
Merck
All Photos(1)

Key Documents

03907

Sigma-Aldrich

Tioconazole

97%

Synonym(s):

1-[2-[(2-Chloro-3-thienyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, Thioconazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13Cl3N2OS
CAS Number:
Molecular Weight:
387.71
EC Number:
MDL number:
UNSPSC Code:
51302325
PubChem Substance ID:
NACRES:
NA.85

Assay

≥96.5% (HPLC)
96.5-103.5% (NT)
97%

form

powder

color

white to off-white

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

SMILES string

Clc1ccc(C(Cn2ccnc2)OCc3ccsc3Cl)c(Cl)c1
Clc1ccc(C(Cn2ccnc2)OCc3ccsc3Cl)c(Cl)c1

InChI

1S/C16H13Cl3N2OS/c17-12-1-2-13(14(18)7-12)15(8-21-5-4-20-10-21)22-9-11-3-6-23-16(11)19/h1-7,10,15H,8-9H2

InChI key

QXHHHPZILQDDPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: pyrimidine

Application

Tioconazole was used to study the synergistic mechanisms of antifungals on Candida albicans as well as significant drug-drug interactions in vivo.

Biochem/physiol Actions

Tioconazole is an antifungal medication of the imidazole class used to treat infections caused by fungus and yeast.It is more active than fluconazole or voriconazole against Candida glabrata mutant strains. It acts by membrane disruption and loss of intracellular ATP.

Packaging

10G

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synergistic action of nikkomycin X/Z with azole antifungals on Candida albicans.
Slawomir Milewski, Fiorenzo Mignini, et. Al
Microbiology, 137, 2155-2161 (1991)
Wenjiang Zhang et al.
Drug metabolism and disposition: the biological fate of chemicals, 30(3), 314-318 (2002-02-21)
The interactions of a panel of antifungal agents with cytochromes P450 (P450s), as a means of predicting potential drug-drug interactions, have not yet been investigated. The objective of this study was to evaluate the specificity and selectivity of five antifungal
A Defontaine et al.
Journal of medical microbiology, 48(7), 663-670 (1999-07-14)
A commercially available disk diffusion procedure was used in a large-scale study to evaluate the susceptibility of a wide range of Candida isolates to polyenes and azoles. With almost all isolates of C. glabrata resistant colonies were present within the
E Ruckenstein et al.
International journal of pharmaceutics, 267(1-2), 121-127 (2003-11-07)
The results obtained previously by Ruckenstein and Shulgin [Int. J. Pharm. 258 (2003a) 193; Int. J. Pharm. 260 (2003b) 283] via the fluctuation theory of solutions regarding the solubility of drugs in binary aqueous mixed solvents were extended in the
Clarisse Zaitz et al.
International journal of dermatology, 45(9), 1071-1073 (2006-09-12)
The present report describes a case of onychoprotothecosis caused by Prototheca wickerhamii in a nonimmunocompromized female. Dermatological examination showed yellowish discoloration of the left and right forefinger nails, showing onicolysis and hyperkeratosis. The repeated isolation of the algae Prototheca (organisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service