Skip to Content
Merck
All Photos(1)

Key Documents

PHR1052

Supelco

Metronidazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

MTZ, 2-Methyl-5-nitroimidazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
Beilstein:
611683
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 603
traceable to Ph. Eur. M1850000
traceable to USP 1442009

API family

metronidazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

159-161 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC1=NC=C([N+]([O-])=O)N1CCO

InChI

1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3

InChI key

VAOCPAMSLUNLGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Metronidazole is a 5-nitroimidazole derivative that is an effective agent against the diseases caused by anaerobic protozoa and bacteria. It is known to exert radio-sensitizing impact on hypoxic tumour cells.

Application

Metronidazole may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0264 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of metronidazole and miconazole in pharmaceutical dosage forms by RP-HPLC
Akay C, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 57(11), 953-957 (2002)
Electrochemical reduction of metronidazole at activated glassy carbon electrode and its determination in pharmaceutical dosage forms
Ozkan SA, et al.
Journal of Pharmaceutical and Biomedical Analysis, 17(2), 299-305 (1998)
Simultaneous determination of miconazole nitrate and metronidazole in different pharmaceutical dosage forms by gas chromatography and flame ionization detector (GC-FID).
Ashour S and Kattan N
International Journal of Biomedical Science : IJBS, 6(1), 13-13 (2010)
Sonja Löfmark et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 50 Suppl 1, S16-S23 (2010-01-14)
Metronidazole has been used for the treatment of infections for >45 years and is still successfully used for the treatment of trichomoniasis, amoebiasis, and giardiasis. Anaerobic bacterial infections caused by Bacteroides species, fusobacteria, and clostridia respond favorably to metronidazole therapy.
T H I Brummer et al.
BJOG : an international journal of obstetrics and gynaecology, 120(10), 1269-1276 (2013-06-22)
To evaluate cefuroxime and metronidazole antibiotic prophylaxis. Observational nonrandomised 1-year prospective cohort study. Fifty-three hospitals in Finland. A total of 5279 women undergoing hysterectomy for benign indications, with cefuroxime given to 4301 and metronidazole given to 2855. Excluding other antibiotics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service