Skip to Content
Merck
All Photos(2)

Key Documents

O3907

Sigma-Aldrich

Octanoic acid

≥98%

Synonym(s):

Acid C8, Caprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747180
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5 (vs air)

Quality Level

vapor pressure

1 mmHg ( 78 °C)

Assay

≥98%

refractive index

n20/D 1.428 (lit.)

bp

237 °C (lit.)

mp

15-17 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC(O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octanoic acid can be used as a reactant:
  • In the SnO/TiO2 catalyzed esterification with polyols to synthesize lubricating esters.
  • To synthesize amides using primary and secondary amines in the presence of a biocatalyst.
It can also be used as a catalyst for oxidizing sulfides into their corresponding sulfoxides and sulfones with H2O2 as the oxidant.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Murata et al.
iScience, 24(9), 102963-102963 (2021-09-02)
Long-chain triglycerides (LCTs) intake strongly stimulates GIP secretion from enteroendocrine K cells and induces obesity and insulin resistance partly due to GIP hypersecretion. In this study, we found that medium-chain triglycerides (MCTs) inhibit GIP secretion after single LCT ingestion and
Palraj Kalimuthu et al.
Analytical chemistry, 92(19), 13110-13117 (2020-09-01)
Proteases are often used as biomarkers of many pathologies as well as of microbial contamination and infection. Therefore, extensive efforts are devoted to the development of protease sensors. Some applications would benefit from wireless monitoring of proteolytic activity at minimal
Elizabeth B Brown et al.
eLife, 10 (2021-05-06)
Chemosensory systems are critical for evaluating the caloric value and potential toxicity of food. While animals can discriminate between thousands of odors, much less is known about the discriminative capabilities of taste systems. Fats and sugars represent calorically potent and
K M Morrill et al.
Journal of dairy science, 95(7), 3987-3996 (2012-06-23)
Our objectives were to evaluate the use of refractometry as a means of estimating immunoglobulin G (IgG) concentration of bovine maternal colostrum (MC) and determine if fractionation of MC using caprylic acid (CA) improved estimates of IgG. Samples (n=85) of
Hong-Fei Tong et al.
Journal of chromatography. A, 1285, 88-96 (2013-03-12)
Hydrophobic charge-induction chromatography (HCIC) is a novel downstream bioprocessing technology for antibody purification and it has several advantages over traditional purification processes. However, its separation selectivity still needs to be improved. In this work, sodium caprylate (NaCA) was used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service