Skip to Content
Merck
All Photos(2)

Documents

32103

Supelco

Posaconazole

VETRANAL®, analytical standard

Synonym(s):

4-[4-[4-[4-[[(3R,5R)-5-(2,4-Difluorophenyl)-5-(1,2,4-triazol-1-ylmethyl)oxolan-3-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-[(2S,3S)-2-hydroxypentan-3-yl]-1,2,4-triazol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H42F2N8O4
CAS Number:
Molecular Weight:
700.78
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC[C@@H]([C@H](C)O)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OC[C@@H]5CO[C@@](C5)(Cn6cncn6)c7ccc(F)cc7F)cc4

InChI

1S/C37H42F2N8O4/c1-3-35(26(2)48)47-36(49)46(25-42-47)31-7-5-29(6-8-31)43-14-16-44(17-15-43)30-9-11-32(12-10-30)50-20-27-19-37(51-21-27,22-45-24-40-23-41-45)33-13-4-28(38)18-34(33)39/h4-13,18,23-27,35,48H,3,14-17,19-22H2,1-2H3/t26-,27+,35-,37-/m0/s1

InChI key

RAGOYPUPXAKGKH-XAKZXMRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Posaconazole is an azole antifungal drug used mostly against zygomycetes.

Application

Posaconazole may have been used as reference standard in HPLC analysis for quantification of posaconazole from human plasma.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu-Ying Yu et al.
Infection and drug resistance, 11, 1537-1547 (2018-10-06)
Trichosporon dohaense is a rare fungal species that has not been described in human invasive infections. In this study, we investigated two T. dohaense isolates from patients with invasive infections in two hospitals in China, as part of the China
Soraya Eugenia Morales-López et al.
Mycopathologia, 181(11-12), 871-878 (2016-09-30)
The presence of the cryptic species belonging to the Candida glabrata complex has not been studied in Argentina. We analyzed a collection of 117 clinical isolates of C. glabrata complex belonging to a National Culture Collection of Instituto Nacional de
Posaconazole as salvage therapy for zygomycosis.
Greenberg, R. N., et al.
Antimicrobial Agents and Chemotherapy, 50.1, 126-133 (2006)
Shuwen Deng et al.
Emerging microbes & infections, 6(12), e109-e109 (2017-12-07)
This study investigated the triazole phenotype and genotypic of clinical Aspergillus fumigatus isolates from China. We determined the triazole susceptibility profiles of 159 A. fumigatus isolates collected between 2011 and 2015 from four different areas in China tested against 10
Dorival Martins et al.
Scientific reports, 9(1), 9185-9185 (2019-06-27)
Fungi respond to antifungal drugs by increasing their antioxidant stress response. How this impacts antifungal efficacy remains controversial and not well understood. Here we examine the role of catalase activity in the resistance of Saccharomyces cerevisiae to the common antifungals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service