Skip to Content
Merck
All Photos(1)

Key Documents

84683

Sigma-Aldrich

Sulfur

purum p.a., ≥99% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99% (T)
form:
powder

vapor density

8.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

grade

purum p.a.

Assay

≥99% (T)

form

powder

autoignition temp.

450 °F

resistivity

2E23 μΩ-cm, 20°C

ign. residue

≤0.2%

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfur is a chalcogen (ore forming element). It is present in abundant quantities in marine and freshwater sediments. It exists in various forms such as pyrite, acid volatile sulfides, elemental sulfur, sulfates and organic sulfur. Quantitative estimation of reduced inorganic sulfur compounds in modern sediments and shales by a chromium reduction technique has been described. Elemental sulfur is employed for the synthesis of arsenic monosulfide and benzothiazole.

Application

Sulfur has been employed as a reactant in the preparation of Cu-doped iron sulfides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Yen Lu et al.
Physical chemistry chemical physics : PCCP, 20(14), 9038-9044 (2018-03-23)
As the feature sizes of devices decrease to the nanoscale, electron microscopy and lithography will become increasingly essential techniques for fabrication and inspection. In this study, we probed the memory effects of MoS2 field-effect transistors (FETs) subjected to electron beam
Shanshan Wang et al.
Nanoscale, 9(35), 13060-13068 (2017-08-25)
We have studied the atomic structure of small secondary domains that nucleate on monolayer MoS
Najmus Saqib et al.
Applied spectroscopy, 71(7), 1593-1599 (2017-02-02)
An optical diagnostic technique to determine the order and concentration of lithium polysulfides in lithium-sulfur (Li-S) battery electrolytes has been developed. One of the major challenges of lithium-sulfur batteries is the problem of polysulfide shuttling between the electrodes, which leads
Xiao-Chen Liu et al.
Science advances, 6(7), eaay4092-eaay4092 (2020-02-29)
Intercalated transition metal dichalcogenides (TMDs) have attracted substantial interest due to their exciting electronic properties. Here, we report a unique approach where copper (Cu) atoms from bulk Cu solid intercalate spontaneously into van der Waals (vdW) gaps of group IV
Ping Wang et al.
ACS applied materials & interfaces, 9(41), 35712-35720 (2017-09-28)
Colloidal semiconductor quantum dots (QDs) have recently emerged as a good candidate for photocatalytic hydrogen (H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service