Skip to Content
Merck
All Photos(3)

Key Documents

414980

Sigma-Aldrich

Sulfur

powder, 99.98% trace metals basis

Synonym(s):

Sulfur powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

Assay

99.98% trace metals basis

form

powder

autoignition temp.

450 °F

resistivity

2E23 μΩ-cm, 20°C

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

density

2.07 g/mL at 25 °C

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfur can be used as a precursor in the sulfurization of molybdenum (Mo) for the preparation of molybdenum disulfide. It can also be used in the formation of a polymeric electrolyte for lithium-sulfur batteries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of different precursors on CVD growth of molybdenum disulfide
Singh A, et al.
Journal of alloys and compounds, 782, 772-779 (2019)
Tuning the morphology and chemical composition of MoS 2 nanostructures
Radovsky G
Journal of materials science & technology, 54(10), 7768-7779 (2019)
Controlling Structural Anisotropy of Anisotropic 2D Layers in Pseudo-1D/2D Material Heterojunctions
Chen B, et al.
Advanced Materials, 29(34), 1701201-1701201 (2017)
Yu Kobayashi et al.
ACS nano, 9(4), 4056-4063 (2015-03-27)
Atomic-layer transition metal dichalcogenides (TMDCs) have attracted appreciable interest due to their tunable band gap, spin-valley physics, and potential device applications. However, the quality of TMDC samples available still poses serious problems, such as inhomogeneous lattice strain, charge doping, and
Gulluzar Bastug et al.
The Journal of organic chemistry, 78(18), 9303-9308 (2013-08-14)
The newly prepared complex [Pd(IPr*(OMe))(cin)(Cl)] provides high catalytic activity for carbon-sulfur cross-coupling reactions. Nonactivated and deactivated aryl halides were successfully coupled with a large variety of aryl- and alkylthiols using this well-defined palladium N-heterocyclic carbene (NHC) complex.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service