Skip to Content
Merck
All Photos(2)

Key Documents

344621

Sigma-Aldrich

Sulfur

flakes, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
NA.23

vapor density

8.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

Assay

≥99.99% trace metals basis

form

flakes

autoignition temp.

450 °F
450 °F

resistivity

2E23 μΩ-cm, 20°C

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

density

2.07 g/mL at 25 °C

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jubok Lee et al.
Nano letters, 21(1), 43-50 (2020-10-15)
The extreme elastic strain of monolayer transition metal dichalcogenides provides an ideal platform to achieve efficient exciton funneling via local strain modulation; however, studies conducted thus far have focused on the use of substrates with fixed strain profiles. We prepared
William Tam et al.
Journal of molecular biology, 425(14), 2450-2462 (2013-04-02)
The assembly of long non-contractile phage tails begins with the formation of the tail tip complex (TTC). TTCs are multi-functional protein structures that mediate host cell adsorption and genome injection. The TTC of phage λ is assembled from multiple copies
Farhad Forouhar et al.
Nature chemical biology, 9(5), 333-338 (2013-04-02)
How living organisms create carbon-sulfur bonds during the biosynthesis of critical sulfur-containing compounds is still poorly understood. The methylthiotransferases MiaB and RimO catalyze sulfur insertion into tRNAs and ribosomal protein S12, respectively. Both belong to a subgroup of radical-S-adenosylmethionine (radical-SAM)
Kuan-Yeow Show et al.
Biotechnology advances, 31(4), 409-420 (2012-12-27)
Biological removal of carbon, nitrogen and sulfur is drawing increasing research interest in search for an efficient and cost-effective wastewater treatment. While extensive work on separate removal of nitrogen and sulfur is well documented, investigation on simultaneous denitrifying sulfide removal
Chemoselective peptide ligation-desulfurization at aspartate.
Robert E Thompson et al.
Angewandte Chemie (International ed. in English), 52(37), 9723-9727 (2013-07-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service