Skip to Content
Merck
All Photos(2)

Key Documents

13825

Sigma-Aldrich

Sulfur

powder, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
NA.55

vapor density

8.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

Assay

≥99.0%

form

powder

autoignition temp.

450 °F

resistivity

2E23 μΩ-cm, 20°C

ash

≤3%

pH

6 (20 °C, 10 g/L)

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfur is present in abundant quantities in marine and freshwater sediments. It exists in the form of various sulfur compounds such as pyrite, acid volatile sulfides, elemental sulfur, sulfates and organic sulfur. Quantitative estimation of reduced inorganic sulfur compounds in modern sediments and shales by a chromium reduction technique has been described.
Sulfur is naturally present in cysteine and methionine amino acids, which can be detected by inductively coupled plasma mass spectrometry(ICP-MS). It is also an important element required for plant growth, since it is useful in lowering the pH of the soil.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfur as the key element for quantitative protein analysis by capillary liquid chromatography coupled to element mass spectrometry
Wind M, et al.
Angewandte Chemie (International Edition in English), 42(29)x`, 3425-3427 (2003)
Mccarty.B.L et al.
Fundamentals of Turfgrass and Agricultural Chemistry (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service