05-1760
Carboxymethylcellulose sodium salt
SAJ first grade
Synonym(s):
Sodium carboxymethylcellulose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
SAJ first grade
form
solid
composition
Na, 6.0-8.5%
availability
available only in Japan
SMILES string
[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C
InChI
1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);
InChI key
DPXJVFZANSGRMM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Field-scale transport and transformation of carboxymethylcellulose-stabilized nano zero-valent iron.
Richard L Johnson et al.
Environmental science & technology, 47(3), 1573-1580 (2013-01-15)
The fate of nano zerovalent iron (nZVI) during subsurface injection was examined using carboxymethylcellulose (CMC) stabilized nZVI in a very large three-dimensional physical model aquifer with detailed monitoring using multiple, complementary detection methods. A fluorescein tracer test in the aquifer
Maintaining skin integrity in patients with incontinent episodes.
Catherine T Milne
Ostomy/wound management, 59(2), 10-10 (2013-03-22)
Zhi-chen Liu et al.
Archives of orthopaedic and trauma surgery, 133(3), 295-301 (2013-01-01)
To evaluate effectiveness of carboxymethylcellulose/polyethylene oxide (CMC/PEO) gel in improving clinical outcomes after the first-time lumbar discectomy. Ninety-three patients with herniated lumbar disc at L4-L5 or L5-S1 were enrolled and randomized into two groups: CMC/PEO gel treatment group and control
Joy Tickle
British journal of community nursing, Suppl, S38-S38 (2013-05-03)
The management of patients with highly-exuding wounds can often lead to the use of unreliable and costly treatments. Patients are frequently found to be at increased risk of infection and delayed healing, which results in a huge negative impact on
M Panca et al.
Journal of wound care, 22(3), 109-110 (2013-05-15)
To estimate the clinical effectiveness and cost effectiveness of using a sodium carboxymethylcellulose dressing (CMC [Aquacel]) and four super absorbent dressings (DryMax Extra[DM], Flivasorb [F], Kerramax [K] and sachet S [S]) in the treatment of highly exuding chronic venous leg
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service