13-1920
Hydrogen peroxide solution
SAJ first grade, ≥30.0%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
Recommended Products
grade
SAJ first grade
vapor pressure
23.3 mmHg ( 30 °C)
form
liquid
reaction suitability
reagent type: oxidant
availability
available only in Japan
concentration
≥30.0%
density
1.13 g/cm3
SMILES string
OO
InChI
1S/H2O2/c1-2/h1-2H
InChI key
MHAJPDPJQMAIIY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stephen J D O'Keefe et al.
Nature communications, 6, 6342-6342 (2015-04-29)
Rates of colon cancer are much higher in African Americans (65:100,000) than in rural South Africans (<5:100,000). The higher rates are associated with higher animal protein and fat, and lower fibre consumption, higher colonic secondary bile acids, lower colonic short-chain
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
Esther P M Tjin et al.
Cancer immunology research, 2(6), 538-546 (2014-06-05)
In this study, we investigated a large series of immune (escape) markers, relevant to T-cell function, as potential biomarkers for clinical outcome following immunotherapy. We retrospectively studied the expression of immune (escape) markers in metastatic melanoma tissues of 27 patients
Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
Osamu Sakai et al.
Investigative ophthalmology & visual science, 56(1), 538-543 (2015-01-13)
The purpose of the present study was to investigate the role of glutathione peroxidase 4 (GPx4) in conjunctival epithelial cells. An immortalized human conjunctival epithelial cell line was used. Cells were transfected with catalase, GPx1, GPx4, SOD1, SOD2, or control
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service