Skip to Content
Merck
All Photos(1)

Key Documents

13-1650

Sigma-Aldrich

Hydrochloric acid

35.0-37.0%, SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Product Name

Hydrochloric acid, SAJ first grade, 35.0-37.0%

grade

SAJ first grade

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

form

aqueous solution

availability

available only in Japan

concentration

35.0-37.0%

bp

>100 °C (lit.)

density

1.18 g/L at 20 °C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc.: This research utilizes hydrochloric acid in the synthesis of zinc-ammonium phosphate from galvanic waste, showcasing its role in resource recovery and waste management (Morgovan et al., 2024).

  • Simple Detection of Pigment Red 53 as a Hazardous Substance in Cosmetic Preparation Using a Polymer Combination of Polystyrene (PS) and Polymethylmethacrylate (PMMA).: Hydrochloric acid is used for the detection of hazardous substances in cosmetics, providing a method for ensuring product safety (Pratiwi et al., 2022).

  • Speciation of tin ions in oxide glass containing iron oxide through solvent extraction and inductively coupled plasma atomic emission spectrometry after the decomposition utilizing ascorbic acid.: This study employs hydrochloric acid in the speciation of tin ions, illustrating its utility in advanced analytical techniques (Saijo et al., 2022).

  • Assessing the behaviour of heavy metals in abandoned phosphogypsum deposits combining electrical resistivity tomography and multivariate analysis.: Hydrochloric acid plays a key role in the analysis of heavy metals in environmental samples, contributing to environmental management and remediation efforts (Vásconez-Maza et al., 2021).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal
Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in
Xuan Zhang et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1365-1375 (2015-04-19)
Intratumoral injection of ultra-small gold nanoparticles (AuNPs) conjugated to doxorubicin (Au-Dox) is effective against both murine B16 and human SK-MEL-28 tumors in mice. Au-Dox suppresses growth of B16 tumors in immunocompetent mice by >70% for at least 19 days. In
Davide R Ceratti et al.
Advanced materials (Deerfield Beach, Fla.), 27(34), 4958-4962 (2015-07-23)
A new, simple bi-phasic dip-coating method is developed. This method is considered as a great improvement of the technique for research, development and production, since expensive, rare, harmful, or time-evolving solutions can now be easily deposited on large surfaces and
Jeroen Aerts et al.
Brain structure & function, 220(5), 2675-2689 (2014-06-25)
Matrix metalloproteinases (MMPs) are Zn(2+)-dependent endopeptidases considered to be essential for normal brain development and neuroplasticity by modulating extracellular matrix proteins, receptors, adhesion molecules, growth factors and cytoskeletal proteins. Specifically, MMP-3 has recently been implicated in synaptic plasticity, hippocampus-dependent learning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service