CLS980016
Pyrex® test tubes, beaded rim
capacity 20 mL
Synonym(s):
Corning® 9800
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
plain neck
round bottom tube
sterility
non-sterile
feature
graduations: no
packaging
case of 576
pack of 72
manufacturer/tradename
Corning 9800-16
O.D. × L
16 mm × 150 mm
capacity
20 mL
color
clear
suitability
suitable for (test)
Looking for similar products? Visit Product Comparison Guide
General description
Beaded rim. Made from special machine-made tubing to give optimum wall thickness. Well annealed, resistant to heat, and chemically stable. Rims are fire-polished. 25 mm tubes do not have marking spot.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service