CLS7080013
Pyrex® VISTA™ test tube
diam. × L 13 mm × 100 mm
Synonym(s):
Corning® 70800
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
neck (Beaded rim)
round bottom tube
sterility
non-sterile
feature
graduations: no
packaging
case of 200
manufacturer/tradename
Corning 70800-13
diam. × L
13 mm × 100 mm
volume
9 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service