Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
test tubes, beaded rim
Synonym(s):
Corning
®
9800
Products
Product Category
tubes (9)
Color
Manufacturer
Capacity (mL)
OD (mm)
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
CLS980016
Pyrex
®
test tubes, beaded rim,
capacity 20 mL
Expand
View Pricing
CLS980025X
Pyrex
®
test tubes, beaded rim,
capacity 70 mL
Expand
View Pricing
CLS980025
Pyrex
®
test tubes, beaded rim,
capacity 55 mL
Expand
View Pricing
CLS980015
Pyrex
®
test tubes, beaded rim,
capacity 14 mL
Expand
View Pricing
CLS980010
Pyrex
®
test tubes, beaded rim,
capacity 3 mL
Expand
View Pricing
CLS980012
Pyrex
®
test tubes, beaded rim,
capacity 5 mL
Expand
View Pricing
CLS980013
Pyrex
®
test tubes, beaded rim,
capacity 9 mL
Expand
View Pricing
CLS980018
Pyrex
®
test tubes, beaded rim,
capacity 27 mL
Expand
View Pricing
CLS980020
Pyrex
®
test tubes, beaded rim,
capacity 34 mL
Expand
View Pricing