Skip to Content
Merck
All Photos(1)

Key Documents

911690

Sigma-Aldrich

C5 Lenalidomide

≥95%

Synonym(s):

3-(5-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione, E3 ligase ligand, Ligand for PROTAC® research

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3O3
CAS Number:
Molecular Weight:
259.26
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

ligand

C5 Lenalidomide

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: ligand

storage temp.

2-8°C

SMILES string

NC1=CC=C2C(CN(C3CCC(NC3=O)=O)C2=O)=C1

InChI

1S/C13H13N3O3/c14-8-1-2-9-7(5-8)6-16(13(9)19)10-3-4-11(17)15-12(10)18/h1-2,5,10H,3-4,6,14H2,(H,15,17,18)

InChI key

WLUIQUZGNPAKRL-UHFFFAOYSA-N

Application

C5 Lenalidomide is a derivative of the Cereblon (CRBN)-binding ligand lenalidomide, which possesses an amine at the C4 position. C5 Lenalidomide can be used to bind the CRBN E3 ubiquitin ligase in targeted protein degradation research or in the synthesis of protein degraders with a C5 exit vector. Learn more about proteolysis-targeting chimeras (PROTACs) and the building blocks to design them in our Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service