904708
Pomalidomide-C6-PEG3-butyl iodide
≥95%
Synonym(s):
N-(2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)-6-(2-(2-((6-iodohexyl)oxy)ethoxy)ethoxy)hexanamide, Crosslinker−E3 Ligase ligand conjugate, Pomalidomide-6-2-2-6-I, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader
About This Item
Recommended Products
ligand
pomalidomide
Assay
≥95%
form
powder or crystals
reaction suitability
reactivity: sulfuryl reactive
reagent type: ligand-linker conjugate
functional group
alkyl halide
storage temp.
2-8°C
SMILES string
ICCCCCCOCCOCCOCCCCCC(NC1=CC=CC2=C1C(N(C3CCC(NC3=O)=O)C2=O)=O)=O
Application
Other Notes
Portal: Building PROTAC® Degraders for Targeted Protein Degradation
Modular PROTAC Design for the Degradation of Oncogenic BCR-ABL
Targeted Protein Degradation by Small Molecules
Small-Molecule PROTACS: New Approaches to Protein Degradation
Targeted Protein Degradation: from Chemical Biology to Drug Discovery
Impact of linker length on the activity of PROTACs
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service