Skip to Content
Merck
All Photos(3)

Documents

T144

Sigma-Aldrich

(±)-Thalidomide

≥98%, powder

Synonym(s):

(±)-2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O4
CAS Number:
Molecular Weight:
258.23
EC Number:
MDL number:
UNSPSC Code:
12352111
eCl@ss:
39180303
PubChem Substance ID:
NACRES:
NA.77

ligand

thalidomide

Assay

≥98%

form

powder

reaction suitability

reagent type: ligand

color

white

solubility

DMSO: 20 mg/mL, clear

originator

Celgene

SMILES string

O=C1CCC(N2C(=O)c3ccccc3C2=O)C(=O)N1

InChI

1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)

InChI key

UEJJHQNACJXSKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thalidomide has been used to study its neuropathological effects in mouse models of Alzheimer′s disease (AD). This study reported that long term administration of thalidomide causes beta-secretase inhibition and subsequently alleviates amyloid-like pathology. Furthermore, thalidomide has also been used to evaluate its teratogenic functions. Thalidomide was found to affect endodermal differentiation and neural development in differentiating human embryonic and induced pluripotent stem cells.

Biochem/physiol Actions

(±)-Thalidomide selectively inhibits biosynthesis of tumor necrosis factor α (TNF-α). It also functions as an inhibitor of angiogenesis, an immunosuppressive agent, a sedative and a teratogen. Furthermore, thalidomide is known to exhibit antitumor functions in refractory multiple myeloma.

Features and Benefits

This compound was developed by Celgene. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Thalidomide is soluble in DMSO at a concentration that is more than 20 mg/ml. It is insoluble in water and ethanol.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoav Mayshar et al.
Journal of cellular and molecular medicine, 15(6), 1393-1401 (2010-06-22)
Teratogens are substances that may cause defects in normal embryonic development while not necessarily being toxic in adults. Identification of possible teratogenic compounds has been historically beset by the species-specific nature of the teratogen response. To examine teratogenic effects on
Marzia Lazzerini et al.
JAMA, 310(20), 2164-2173 (2013-11-28)
Pediatric-onset Crohn disease is more aggressive than adult-onset disease, has high rates of resistance to existing drugs, and can lead to permanent impairments. Few trials have evaluated new drugs for refractory Crohn disease in children. To determine whether thalidomide is
S Singhal et al.
The New England journal of medicine, 341(21), 1565-1571 (1999-11-24)
Patients with myeloma who relapse after high-dose chemotherapy have few therapeutic options. Since increased bone marrow vascularity imparts a poor prognosis in myeloma, we evaluated the efficacy of thalidomide, which has antiangiogenic properties, in patients with refractory disease. Eighty-four previously
Karin Lauschke et al.
Archives of toxicology, 95(5), 1659-1670 (2021-03-05)
To test large numbers of chemicals for developmental toxicity, rapid in vitro tests with standardized readouts for automated data acquisition are needed. However, the most widely used assay, the embryonic stem cell test, relies on the counting of beating embryoid
Yang Yang et al.
Pharmaceutics, 15(4) (2023-04-28)
Thalidomide (THD), a synthetic derivative of glutamic acid, was initially used as a sedative and antiemetic until the 1960s, when it was found to cause devastating teratogenic effects. However, subsequent studies have clearly demonstrated the anti-inflammatory, anti-angiogenic, and immunomodulatory properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service