911704
C5 Lenalidomine-C3-NH2 hydrochloride
≥95%
Synonym(s):
4-Amino-N-(2-(2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-5-yl)butanamide hydrochloride, C5 Lenalidomide conjugate, Crosslinker-E3 Ligase ligand conjugate, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader
About This Item
Recommended Products
ligand
C5 Lenalidomide
Quality Level
Assay
≥95%
form
powder or crystals
reaction suitability
reactivity: carboxyl reactive
reagent type: ligand-linker conjugate
functional group
amine
storage temp.
2-8°C
SMILES string
O=C1N(C2CCC(NC2=O)=O)CC3=CC(NC(CCCN)=O)=CC=C31.Cl
InChI key
OMSXFPPNCRZPJX-UHFFFAOYSA-N
Related Categories
Application
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - STOT RE 2
Target Organs
Blood
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service