Skip to Content
Merck
All Photos(2)

Key Documents

900549

Sigma-Aldrich

Poly(3-hexylthiophene-2,5-diyl)

regioregular, average Mw 85,000-100,000

Synonym(s):

P3HT, P3HT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H14S)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

Metal purity: ≤Mg: 100 ppm
≤Ni: 100 ppm

Regioregularity: ≥90%

Quality Level

form

solid

mol wt

average Mw 85,000-100,000

InChI

1S/C12H20S/c1-4-5-6-7-8-12-9-10(2)13-11(12)3/h9H,4-8H2,1-3H3

InChI key

DUFPJSOXRHVDOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service