Skip to Content
Merck
All Photos(2)

Key Documents

450650

Sigma-Aldrich

Poly(3-dodecylthiophene-2,5-diyl)

regioregular, average Mw ~60,000

Synonym(s):

P3DDT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

mol wt

average Mw ~60,000

solubility

chloroform: soluble

fluorescence

λex 442 nm; λem 568 nm in chloroform

OPV Device Performance

ITO/PEDOT:PSS/P3DDT/PC61BM (1:3)/LiF/Al

  • Short-circuit current density (Jsc): 2.9 mA/cm2
  • Open-circuit voltage (Voc): 0.6 V
  • Fill Factor (FF): 0.38
  • Power Conversion Efficiency (PCE): 0.65 %

semiconductor properties

P-type (mobility=1E-4-1E-1 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

General description

Conducting Polymer. Greater than 98.5% head-to-tail regiospecific conformation.
Forms a shiny red-purple film.

Application

Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
Conducting polymer.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.

Legal Information

Product of Rieke® Metals, Inc. ®Reg. trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of Annealing on the Nanomorphology and Performance of Poly(alkylthiophene):Fullerene Bulk-Heterojunction Solar Cells
Nguyen, L. H.; et al.
Advances in Functional Materials, 17, 1071-1078 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service