Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(3-hexylthiophene-2,5-diyl)
Synonym(s):
P3HT
Linear Formula:
(C
10
H
14
S)
n
CAS Number:
104934-50-1
Products
Brand
Sigma-Aldrich (4)
Sustainability
Feature
Melting Point (°C)
Physical Form
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
445703
Poly(3-hexylthiophene-2,5-diyl),
regioregular
Expand
View Pricing
900550
Poly(3-hexylthiophene-2,5-diyl),
regioregular, average M
w
50,000-75,000
Expand
View Pricing
900563
Poly(3-hexylthiophene-2,5-diyl),
regioregular, average M
w
20,000-45,000
Expand
View Pricing
900549
Poly(3-hexylthiophene-2,5-diyl),
regioregular, average M
w
85,000-100,000
Expand
View Pricing