Skip to Content
Merck
All Photos(3)

Key Documents

900550

Sigma-Aldrich

Poly(3-hexylthiophene-2,5-diyl)

regioregular, average Mw 50,000-75,000

Synonym(s):

P3HT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H14S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

Metal purity: Mg: ≤100 ppm
Ni: ≤100 ppm

Regioregularity: ≥90%

Quality Level

form

solid

mol wt

average Mw 50,000-75,000

SMILES string

[s]1c(c(cc1C)CCCCCC)C

InChI

1S/C12H20S/c1-4-5-6-7-8-12-9-10(2)13-11(12)3/h9H,4-8H2,1-3H3

InChI key

DUFPJSOXRHVDOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-hexylthiophene-2,5-diyl) (P3HT) is a polymer with a simple chemical structure, high chemical stability, good semiconducting properties, spectral absorption matching solar spectrum, controlled synthetic method, and wide commercial availability. It is widely used as a donor material in organic solar cells.

Application

Poly(3-hexylthiophene-2,5-diyl) can be used as a semiconductor material to fabricate high-performance organic field-effect transistors(OFETs). It can also be used to prepare highly sensitive pressure and temperature-sensitive sensors for bio-signal monitoring.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service