Skip to Content
Merck
All Photos(1)

Key Documents

797391

Sigma-Aldrich

D131 dye

>95% (HPLC)

Synonym(s):

2-Cyano-3-[4-[4-(2,2-diphenylethenyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopent[b]indol-7-yl]-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H28N2O2
Molecular Weight:
508.61
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

>95% (HPLC)

form

solid

solubility

DMF: soluble
DMSO: soluble
THF: soluble
acetone: soluble
chloroform: soluble
ethanol: soluble
methanol: soluble
water: insoluble

λmax

425 nm in methanol

storage temp.

2-8°C

SMILES string

OC(/C(C#N)=C/C(C=C1)=CC2=C1N(C3=CC=C(C=C(C4=CC=CC=C4)C5=CC=CC=C5)C=C3)C6C2CCC6)=O

InChI

1S/C35H28N2O2/c36-23-28(35(38)39)20-25-16-19-34-32(22-25)30-12-7-13-33(30)37(34)29-17-14-24(15-18-29)21-31(26-8-3-1-4-9-26)27-10-5-2-6-11-27/h1-6,8-11,14-22,30,33H,7,12-13H2,(H,38,39)/b28-20+

InChI key

GOTRYMLNXIJMCB-VFCFBJKWSA-N

General description

D131 dye is an indoline based organic dye that has a large band-gap and can be used as a sensitizer in organic electronics. It shows a power conversion efficiency of 5.6% and can enhance photo-excitation of the electrochemical devices.

Application

D131 can be used in dye sensitized solar cells (DSSCs) as a photosensitizer with high charge mobility and absorption coefficient.
Organic photosensitive yellow dye having excellent photoelectric conversion efficiency as a metal-free dye,
Significant improvement in the conversion efficiency (11.1%) of black-dye-based dye-sensitized solar cells by cosensitization with D131 dye

Legal Information

Product of Mitsubishi Paper Mills; Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Significant improvement in the conversion efficiency of black-dye-baseddye-sensitized solar cells by cosensitization with organic dye
Hironobu Ozawa; Ryosuke Shimizu and Hironori Arakawa
Royal Society of Chemistry Advances, 2, 3198-3200 (2012)
Indoline dyes with various acceptors for dye-sensitized solar cells
Kim JY, et al.
Current Applied Physics, 11(1), S117-S121 (2011)
Development of Flexible Dye-sensitized Solar Cell Based on Predyed Zinc Oxide Nanoparticle
Zhang L and Konno A
International Journal of Electrochemical Science, 13(1), 344-352 (2018)
A selective co-sensitization approach to increase photon conversion efficiency and electron lifetime in dye-sensitized solar cells
Nguyen LH, et al.
Physical Chemistry Chemical Physics, 14(47), 16182-16186 (2012)
Panchromatic engineering for dye-sensitized solar cells
Jun-Ho Yum; Etienne Baranoff; Sophie Wenger; et al.
Energy & Environmental Science, 4, 842-842 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service