Skip to Content
Merck
All Photos(1)

Key Documents

745944

Sigma-Aldrich

D102 Dye

95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H30N2O3S2
CAS Number:
Molecular Weight:
614.78
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95% (HPLC)

form

powder

mp

238-243 °C

λmax

499 nm in DMF

SMILES string

S=C(S/C1=C\C(C=C2)=CC3=C2N(C4=CC=C(C=C(C5=CC=CC=C5)C6=CC=CC=C6)C=C4)C7C3CCC7)N(CC(O)=O)C1=O

InChI

1S/C37H30N2O3S2/c40-35(41)23-38-36(42)34(44-37(38)43)22-25-16-19-33-31(21-25)29-12-7-13-32(29)39(33)28-17-14-24(15-18-28)20-30(26-8-3-1-4-9-26)27-10-5-2-6-11-27/h1-6,8-11,14-22,29,32H,7,12-13,23H2,(H,40,41)/b34-22-

InChI key

XGMCROHUTRXETK-VQNDASPWSA-N

General description

D102 Dye is a low cost indoline based organic dye that can be used as a photosensitizer. Its power conversion efficiency is over 4% with a strong absorption coefficient (55800 Lmol-1cm-1 at 490nm). It has a high extinction coefficient in comparison to the ruthenium dye. It can improve the performance of electrochemical devices.

Application

D102 Dye can be used as a sensitizing material that can be coated on the semiconducting electrode for the fabrication of metal-free dye sensitized solar cells.
Photosensitive Red dye having excellent photoelectric conversion efficiency as a metal-free dye. Efficiency of DSSC device - η > 5.4%
Cell area: 0.25 cm2 (0.5 cm × 0.5 cm)
TiO2 Thickness: 5 μm
TiO2: screen printing
Electrolyte: 0.1 M LiI; 0.05 M I2; 0.6 M Dimethyl propylimidazolium
Iodide; 0.05 M t-Butylpyridine in 3-Methoxypropionitrile
Light source: AM1.5
Counter Electrode: Pt/Cr spatter glass
Dye adsorption: 30 °C/ in 3 hr. (t-BuOH/acetonitrile soln.)

Legal Information

Product of Mitsubishi Paper Mills; Ltd

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic dye-sensitized ionic liquid based solar cells: remarkable enhancement in performance through molecular design of indoline sensitizers
Kuang D, et al.
Angewandte Chemie (International Edition in English), 120(10), 1949-1953 (2008)
Dye molecules for simple co-sensitization process: fabrication of mixed-dye-sensitized solar cells.
Mutsumi Kimura et al.
Angewandte Chemie (International ed. in English), 51(18), 4371-4374 (2012-02-10)
Highly efficient poly (3-hexylthiophene) based monolithic dye-sensitized solar cells with carbon counter electrode
Wang H, et al.
Energy & Environmental Science, 4(6), 2025-2029 (2011)
Metal-free indoline dye sensitized solar cells based on nanocrystalline Zn2SnO4
Huang L, et al.
Electrochemical Communications, 12(2), 319-322 (2010)
Highly efficient nanoporous TiO2-polythiophene hybrid solar cells based on interfacial modification using a metal-free organic dye
Zhu R, et al.
Advanced Materials, 21(9), 994-1000 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service