Skip to Content
Merck
All Photos(1)

Key Documents

791423

Sigma-Aldrich

C101 Dye

Synonym(s):

cis-Bis(isothiocyanato)(2,2′-bipyridyl-4,4′-dicarboxylato)(4,4′-bis(5-hexylthiophen-2-yl)-2,2′-bipyridyl)ruthenium(II), Greatcell Solar®, Ruthenate(2−), [[2,2′-bipyridine]-4,4′-dicarboxylato(2−)-N1,N1′][4,4′-bis(5-hexyl-2-thienyl)-2,2′-bipyridine-N1,N1′]bis(thiocyanato-N)-, hydrogen (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H44N6O4RuS4
CAS Number:
Molecular Weight:
950.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder (maroon)

Quality Level

λmax

546, 403, 338, 307 nm (lit.)

SMILES string

OC(C1=CC(C2=CC(C(O)=O)=CC=N2)=NC=C1)=O.S=C=N[Ru]N=C=S.CCCCCCC(S3)=CC=C3C4=CC=NC(C5=NC=CC(C6=CC=C(CCCCCC)S6)=C5)=C4

InChI

1S/C30H36N2S2.C12H8N2O4.2CNS.Ru/c1-3-5-7-9-11-25-13-15-29(33-25)23-17-19-31-27(21-23)28-22-24(18-20-32-28)30-16-14-26(34-30)12-10-8-6-4-2;15-11(16)7-1-3-13-9(5-7)10-6-8(12(17)18)2-4-14-10;2*2-1-3;/h13-22H,3-12H2,1-2H3;1-6H,(H,15,16)(H,17,18);;;/q;;2*-1;+2

InChI key

BXXRBPUFQSDMHJ-UHFFFAOYSA-N

General description

C101 Dye is ruthenium (Ru) based organic dye that can be used as a heteroleptic thiophene-based sensitizer. It has a high extinction coefficient of 17500 Lmol-1cm-1 and facilitates the enhancement of optoelectronic properties of electrochemical devices.

Application

A ruthenium sensitizer with high molar extinction coefficient for high-performance dye-sensitized solar cells
Along with an acetonitrile-based electrolyte; the C101 sensitizer achieved a high efficeincy of 11.0-11.3% in the dye-sensitized solar cells under the illumination of AM 1.5G full sunlight.
C101 Dye can be used as a sensitizer for the fabrication of dye sensitized solar cells (DSSCs) with a power conversion efficiency (PCE) of 10−11%.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dye-sensitized solar cells employing a single film of mesoporous TiO2 beads achieve power conversion efficiencies over 10%
Sauvage F, et al.
ACS Nano, 4(8), 4420-4425 (2010)
Hierarchical TiO2 photoanode for dye-sensitized solar cells
Sauvage F, et al.
Nano Letters, 10(7), 2562-2567 (2010)
Dye-sensitized solar cells with a high absorptivity ruthenium sensitizer featuring a 2-(hexylthio) thiophene conjugated bipyridine
Cao Y, et al.
The Journal of Physical Chemistry C, 113(15), 6290-6297 (2009)
Dye-sensitized solar cells with inkjet-printed dyes
Hashmi SG, et al.
Energy & Environmental Science, 9(7), 2453-2462 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service