Skip to Content
Merck
All Photos(1)

Documents

323756

Sigma-Aldrich

Merocyanine 540

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N3NaO6S2
CAS Number:
Molecular Weight:
569.67
Beilstein:
3585252
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

composition

Dye content, 90%

mp

285 °C (dec.) (lit.)

λmax

555 nm

fluorescence

λex 555 nm; λem 578 nm in methanol

SMILES string

[Na+].CCCCN1C(=O)C(=C\C=C\C=C2/Oc3ccccc3N2CCCS([O-])(=O)=O)\C(=O)N(CCCC)C1=S

InChI

1S/C26H33N3O6S2.Na/c1-3-5-16-28-24(30)20(25(31)29(26(28)36)17-6-4-2)12-7-10-15-23-27(18-11-19-37(32,33)34)21-13-8-9-14-22(21)35-23;/h7-10,12-15H,3-6,11,16-19H2,1-2H3,(H,32,33,34);/q;+1/p-1/b10-7+,23-15-;

InChI key

OSQUFVVXNRMSHL-LTHRDKTGSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent axonal stain.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

B Fernandez-Fuertes et al.
Reproduction (Cambridge, England), 155(6), 481-492 (2018-04-06)
This study tested the hypothesis that sperm sialic acid (Sia) is required to reach the site of fertilization, and that successful fertilization requires recognition of Sia from both the sperm and oocyte to occur. In addition, it has recently been
Graciela Quintana Saalfeld et al.
Environmental science and pollution research international, 25(3), 2924-2931 (2017-11-18)
Reproductive effects caused by the exposure to environmentally relevant dosages of atrazine on wild animals are poorly understood. This study evaluated the effects of three dosages of atrazine on sperm parameters of adult Calomys laucha males. Adult mice were orally
Maria J Martinez-Alborcia et al.
PloS one, 7(5), e36550-e36550 (2012-05-09)
In the present study, the impact of non-functional spermatozoa on the cryopreservation success of functional boar spermatozoa was evaluated. Fifteen sperm-rich ejaculate fractions collected from five fertile boars were frozen with different proportions of induced non-functional sperm (0--native semen sample-
K R Pool et al.
Domestic animal endocrinology, 74, 106516-106516 (2020-07-28)
The ability of the neurohormone melatonin to ameliorate cryopreservation-induced damage to spermatozoa has been demonstrated in several domestic species. However, it is unclear how these protective effects are conferred, with improvements in sperm quality ambiguously attributed to the general antioxidant
Ana M Adamo et al.
Neurotoxicity research, 17(1), 1-14 (2009-09-29)
Although a requirement of zinc (Zn) for normal brain development is well documented, the extent to which Zn can modulate neuronal proliferation and apoptosis is not clear. Thus, we investigated the role of Zn in the regulation of these two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service