Skip to Content
Merck
All Photos(2)

Documents

765155

Sigma-Aldrich

Lithium titanate

greener alternative

spinel, electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

LTO, Lithium titanate spinel oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li4Ti5O12
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

description

Nominal Voltage: 1.5 V, Li/Li+

Assay

≥98%

form

sheet

composition

loading, ≥80%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

≥80% loading

size

5 in. × 10 in.

thickness

25-50 μm

particle size

1.5-3 μm (typical)

capacity

150 mAh/g(minimum)
160 mAh/g(nominal at 0.1C)

mp

>1000 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[Li+].[Li+].[Li+].[O-][Ti](=O)O[Ti](O[Ti]([O-])=O)(O[Ti]([O-])=O)O[Ti]([O-])=O

InChI

1S/4Li.12O.5Ti/q4*+1;;;;;;;;;4*-1;;;;;

InChI key

BNQVSKURWGZJMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium titanate (LTO) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium titanate (LTO) spinel electrode sheet can be used as an anode material, which shows an ion conductivity of 10−3 Scm−1 at room temperature. It can also be used as an alternative to conventional graphite materials. It can further be used in the fabrication of high-performance lithium-ion batteries for electric vehicles (EVs).

Other Notes

Crystal Structure: Spinel, cubic, Fd-3m

Operating Condiditons:
  • Recommended maximum charge voltage: 3.0 V vs Li/Li+
  • Recommended maximum charge current: 5 C
  • Recommended cut-off voltage for discharge: 1.0 V vs Li/Li+
  • Recommended maximum discharge current: 10 C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wu, H-C.; et al.
Journal of Power Sources, 197, 301-301 (2012)
Jansen, A. N.; et al.
Journal of Power Sources, 902, 81-82 (1999)
Ionic conductivity, lithium insertion and extraction of lanthanum lithium titanate
Chen CH and Amine K
Solid State Ionics, 144(1-2), 51-57 (2001)
Lithium titanate tailored by cathodically induced graphene for an ultrafast lithium ion battery
Yang Y, et al.
Advances in Functional Materials, 24(27), 4349-4356 (2014)
Electrochemical properties of doped lithium titanate compounds and their performance in lithium rechargeable batteries
Shenouda AY and Murali KR
Journal of Power Sources, 176(1), 332-339 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service