Skip to Content
Merck
All Photos(2)

Key Documents

765171

Sigma-Aldrich

Lithium nickel cobalt aluminium oxide

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

NCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNi0.8Co0.15Al0.05O2
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

description

Nominal Voltage: 3.7 V, Li/Li+

Assay

≥98%

composition

loading, ≥80%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

≥80% loading

size

5 in. × 10 in.

thickness

12-25 μm

particle size

10-13 μm (typical)

capacity

150 mAh/g(minimum)
180 mAh/g(nominal at 0.1C)

mp

>1000 °C

application(s)

battery manufacturing

greener alternative category

General description

Lithium nickel cobalt aluminium oxide (NCA) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

NCA is Aluminum doped Lithium nickel cobalt oxide (LNCO). Al doping is found very effective to suppress the cell impedance rise by stabilizing the charge-transfer impedance on the cathode side besides increasing the thermal stability of the material. NCA shows excellent electrochemical performance.

The NCA casted electrode sheets can be cut into appropriate size and is ready to be used in lithium ion batteries.

Other Notes

Crystal Structure: Rhombohedral

Operating Condiditons:
  • Recommended maximum charge voltage: 4.3 V vs Li/Li+
  • Recommended maximum charge current: 4C
  • Recommended cut-off voltage for discharge: 3.0 V vs Li/Li+
  • Recommended charge method: constant current - constant voltage

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen, C. H.; et al.
Journal of Power Sources, 128, 278-278 (2004)
Tran, H. Y.; et al.
Journal of the Electrochemical Society, 158, A556-A556 (2011)
The Li-ion rechargeable battery: a perspective
Goodenough JB and Park K
Journal of the American Chemical Society, 135(4), 1167-1176 (2013)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service