Skip to Content
Merck
All Photos(5)

Key Documents

761001

Sigma-Aldrich

Lithium nickel manganese cobalt oxide

greener alternative

powder, <0.5 μm particle size, >98%

Synonym(s):

NMC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNi0.33Mn0.33Co0.33O2
CAS Number:
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

Assay

>98%

form

powder

mol wt

Mw 96.46 g/mol

composition

LiNi0.33Mn0.33Co0.33O2

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<0.5 μm

mp

>290 °C (lit.)

density

2.11 g/cm3

application(s)

battery manufacturing

greener alternative category

General description

Lithium nickel manganese cobalt oxide (NMC) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

NMC is a novel lithium insertion electrode material for advanced lithium-ion batteries. Mn doping significantly increases the thermal stability besides increasing the electrochemical charge-discharge behavior.
NMC111 (lithium nickel-manganese-cobalt oxide with a stoichiometry of 1:1:1) is a promising cathode material used in advanced lithium-ion batteries, particularly for electric vehicle applications, due to its high energy density and long cycle life. NMC111 powder has a layered crystal structure that enables efficient, reversible lithium-ion diffusion, which is essential for good electrochemical performance. Manganese doping improves the thermal stability of the material while enhancing its electrochemical charge-discharge behavior. Compared to other NMC materials, NMC111 has a higher thermal stability and is less prone to thermal runaway, making it a safer option for battery applications.

Features and Benefits

Our NMC111 has engineered improvements over other LiNMC materials including thermal stability and high conductivity:
  • More Reliable
  • Longer Cycle Life
  • Bulk and Pilot Scale Available

Legal Information

Product of Engi-Mat Co.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A bi-functional lithium difluoro (oxalato) borate additive for lithium cobalt oxide/lithium nickel manganese cobalt oxide cathodes and silicon/graphite anodes in lithium-ion batteries at elevated temperatures
Lee SJ, et al.
Electrochimica Acta, 137, 1-8 (2014)
Belharouak, I.; et al.
Journal of Power Sources, 123, 247-247 (2003)
Impedance change and capacity fade of lithium nickel manganese cobalt oxide-based batteries during calendar aging
Schmitt J, et al.
Journal of Power Sources, 353, 183-194 (2017)
Choi, J.; Manthiram, A.; J.
Journal of the Electrochemical Society, 152, A1714-A1714 (2005)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service