Skip to Content
Merck
All Photos(1)

Key Documents

736473

Sigma-Aldrich

Silver, dispersion

nanoparticle, 30-35 wt. % in triethylene glycol monoethyl ether, spec. resistivity ~ 7 μΩ-cm, for printing on ITO films

Synonym(s):

Silverjet DGP-40TE-20C, silver ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

description

surface tension 35 - 40 dyn/cm

form

dispersion
nanoparticle

concentration

30-35 wt. % in triethylene glycol monoethyl ether

refractive index

n20/D 1.333

particle size

≤50 nm

viscosity

10-18 cP

density

1.45 g/mL±0.05 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver is a good conductor and is resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles). Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.

Curing Temperature: 180-200 °C

Recommended substrates: ITO film, Ceramic layer

Recommended washing solvent: Polar Solvents: Ethyl Alcohol, IPA etc.

Legal Information

Product of ANP, Inc.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative Study of Loading of Anodic Porous Alumina with Silver Nanoparticles Using Different Method.
Thorat S, et al.
Materials, 6, 206-216 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service