736511
Silver, dispersion
nanoparticle, 40-50 wt. %, spec. resistivity < 0.001 Ω-cm, for printing on ITO and glass
Synonym(s):
Silverjet DGH-55HTG, silver ink
About This Item
Recommended Products
description
surface tension 40 - 70 mN m-1
form
dispersion
nanoparticle
concentration
40-50 wt. %
refractive index
n20/D 1.333
particle size
≤10 nm
viscosity
900-1600 mPa.s ((25°C))
density
1.5-1.8 g/mL at 25 °C
storage temp.
20-25°C
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
217.9 °F
Flash Point(C)
103.3 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service