Skip to Content
Merck
All Photos(4)

Documents

798738

Sigma-Aldrich

Silver nanoparticle ink

30 wt % dispersion in ethylene glycol

Synonym(s):

Ag ink, Conductive ink, Silver ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

specific gravity 1.53 +/- 0.05 g/ml

Quality Level

form

suspension

concentration

30 wt % dispersion in ethylene glycol

resistivity

≤20 μΩ-cm (after sintering at 200 °C, 2h)
≤45 μΩ-cm (after sintering at 120 °C, 2h)
≤7 μΩ-cm (after sintering at 240 °C, 4h)

particle size

115 nm (d90 (by Brookhaven))
70 nm (d50 (by Brookhaven))

avg. part. size

180 nm

surface tension

45 dyn/cm

viscosity

28 cP(25 °C, Brookfield) (Con Spindle 42)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver is a good conductor and resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles). Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.

Application

The ink is suitable for digital printing such as Inkjet and Aerosol systems, Photovoltaic Cells Printed Electronics, FPD, RFID, PCB Decoration.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanostructured Materials Through Ultrasonic Spray Pyrolysis
Bang JH, et al.
Material Matters, 7(2) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service