Skip to Content
Merck
All Photos(3)

Key Documents

730815

Sigma-Aldrich

Silver, dispersion

nanoparticles, 60 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

nanoparticles

Quality Level

contains

sodium citrate as stabilizer

concentration

0.02 mg/mL in aqueous buffer

refractive index

n20/D 1.333

particle size

60 nm (TEM)

density

0.9976 g/mL at 25 °C

fluorescence

λem 431 nm FWHM 81 nm

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo Rodrigues da Conceição et al.
Metabolic brain disease, 34(3), 705-713 (2019-02-01)
Silver nanoparticles (AgNPs) are clusters of silver atoms with diameters that range from 1 to 100 nm. Due to the various shapes and large surface areas, AgNPs have been employed in the food and textile industries and medical fields. Therefore, because

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service